5-(3-Methylphenyl)-1,3,4-oxadiazol-2-amine 95%
Supplier: Apollo Scientific
OR946700-1G
OR946700-250MG
APOSOR946700-1GEA
350
EUR
APOSOR946700-1G
APOSOR946700-250MG
5-(3-Methylphenyl)-1,3,4-oxadiazol-2-amine 95%
5-(3-Methylphenyl)-1,3,4-oxadiazol-2-amine
Formula:
C₉H₉N₃O MW: 175,187 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00469195 CAS Number: 109060-64-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...