3-Chloro-4-ethoxybenzonitrile 95%

Supplier: Apollo Scientific

OR947166-1G OR947166-250MG
APOSOR947166-1GEA 771 EUR
APOSOR947166-1G APOSOR947166-250MG
3-Chloro-4-ethoxybenzonitrile 95%
3-Chloro-4-ethoxybenzonitrile
Formula: C₉H₈ClNO
MW: 181,619 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10666511
CAS Number: 916596-02-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR