1-Chloro-4-methoxyisoquinoline 95%
Supplier: Apollo Scientific
OR947329-1G
OR947329-250MG
APOSOR947329-1GEA
252
EUR
APOSOR947329-1G
APOSOR947329-250MG
1-Chloro-4-methoxyisoquinoline 95%
1-Chloro-4-methoxyisoquinoline
Formula:
C₁₀H₈ClNO MW: 193,63 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11846289 CAS Number: 3336-60-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...