6-Bromo-2-oxaspiro[3.3]heptane
Supplier: Apollo Scientific
OR947407-1G
OR947407-250MG
OR947407-500MG
APOSOR947407-1GEA
1020
EUR
APOSOR947407-1G
APOSOR947407-250MG
APOSOR947407-500MG
6-Bromo-2-oxaspiro[3.3]heptane
6-Bromo-2-oxaspiro[3.3]heptane
Formula:
C₆H₉BrO MW: 177,039 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD22419347 CAS Number: 1363380-78-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...