5-Methyl-6-nitro-1H-indazole 95%

Supplier: Apollo Scientific

OR948085-5G
APOSOR948085-5GEA 0 EUR
APOSOR948085-5G
5-Methyl-6-nitro-1H-indazole 95%
5-Methyl-6-nitro-1H-indazole
Formula: C₈H₇N₃O₂
MW: 177,16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09026996
CAS Number: 72521-00-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR