Ethyl d-homoalaninate hydrochloride 95%
Supplier: Apollo Scientific
OR948168-1G
OR948168-5G
APOSOR948168-1GEA
174
EUR
APOSOR948168-1G
APOSOR948168-5G
Ethyl d-homoalaninate hydrochloride 95%
Ethyl d-homoalaninate hydrochloride
Formula:
C₆H₁₃NO₂·ClH Storage Temperature: Ambient |
MDL Number:
MFCD11848831 CAS Number: 127641-80-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...