b-Amino-4-methoxy-benzeneethanol 97%

Supplier: Apollo Scientific

Synonyms: 2-Amino-2-(4-methoxyphenyl)ethanol

OR948217-1G OR948217-250MG
APOSOR948217-1GEA 393 EUR
APOSOR948217-1G APOSOR948217-250MG
b-Amino-4-methoxy-benzeneethanol 97%
b-Amino-4-methoxy-benzeneethanol
Formula: C₉H₁₃NO₂
MW: 167,205 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08752776
CAS Number: 138713-55-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR