5-Chloro-7-nitro-1H-indole 95%

Supplier: Apollo Scientific

OR948278-100MG OR948278-1G OR948278-250MG
APOSOR948278-100MGEA 50.9 EUR
APOSOR948278-100MG APOSOR948278-1G APOSOR948278-250MG
5-Chloro-7-nitro-1H-indole 95%
5-Chloro-7-nitro-1H-indole
Formula: C₈H₅ClN₂O₂
Storage Temperature: Ambient
MDL Number: MFCD20923231
CAS Number: 1197181-29-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR