5-(Chloromethyl)-2-methyl-1,3-thiazole 95%

Supplier: Apollo Scientific

Synonyms: 5-(Chloromethyl)-2-methylthiazole

OR949632-1G OR949632-250MG
APOSOR949632-1GEA 487 EUR
APOSOR949632-1G APOSOR949632-250MG
5-(Chloromethyl)-2-methyl-1,3-thiazole 95%
5-(Chloromethyl)-2-methyl-1,3-thiazole
Formula: C₅H₆ClNS
MW: 147,626 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11111716
CAS Number: 63140-11-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR