3H-Imidazo[4,5-b]pyridine-5-carboxylic acid 95%
Supplier: Apollo Scientific
OR949643-1G
OR949643-5G
APOSOR949643-1GEA
37.7
EUR
APOSOR949643-1G
APOSOR949643-5G
3H-Imidazo[4,5-b]pyridine-5-carboxylic acid 95%
3H-Imidazo[4,5-b]pyridine-5-carboxylic acid
Formula:
C₇H₅N₃O₂ MW: 163,134 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09863858 CAS Number: 1019108-05-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...