4-Chloro-2-methoxynicotinic acid 95%

Supplier: Apollo Scientific

OR949736-1G OR949736-250MG
APOSOR949736-1GEA 68.4 EUR
APOSOR949736-1G APOSOR949736-250MG
4-Chloro-2-methoxynicotinic acid 95%
4-Chloro-2-methoxynicotinic acid
Formula: C₇H₆ClNO₃
MW: 187,58 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13188728
CAS Number: 605661-81-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR