1,4-Diisopropoxybenzene 95%
Supplier: Apollo Scientific
OR949969-1G
OR949969-5G
APOSOR949969-1GEA
319
EUR
APOSOR949969-1G
APOSOR949969-5G
1,4-Diisopropoxybenzene 95%
1,4-Diisopropoxybenzene
Formula:
C₁₂H₁₈O₂ Storage Temperature: Ambient |
MDL Number:
MFCD00128849 CAS Number: 7495-78-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...