2-Ethyl-[1,3,4]oxadiazole ≥98%

Supplier: Apollo Scientific

Synonyms: 2-Ethyl-1,3,4-oxadiazole

OR950126-10G OR950126-1G OR950126-5G
APOSOR950126-10GEA 2170 EUR
APOSOR950126-10G APOSOR950126-1G APOSOR950126-5G
2-Ethyl-[1,3,4]oxadiazole ≥98%
2-Ethyl-[1,3,4]oxadiazole
Formula: C₄H₆N₂O
MW: 98,103 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09755521
CAS Number: 13148-61-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR