2-(Allyloxy)phenol 95%

Supplier: Apollo Scientific

OR950199-100MG OR950199-1G OR950199-250MG
APOSOR950199-100MGEA 205 EUR
APOSOR950199-100MG APOSOR950199-1G APOSOR950199-250MG
2-(Allyloxy)phenol 95%
2-(Allyloxy)phenol
Formula: C₉H₁₀O₂
MW: 150,175 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00072617
CAS Number: 1126-20-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR