4-Chloro-6-methyl-1H-indazole ≥98%
Supplier: Apollo Scientific
OR950400-1G
OR950400-250MG
APOSOR950400-1GEA
623
EUR
APOSOR950400-1G
APOSOR950400-250MG
4-Chloro-6-methyl-1H-indazole ≥98%
4-Chloro-6-methyl-1H-indazole
Formula:
C₈H₇ClN₂ MW: 166.608 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07781503 CAS Number: 885521-74-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...