1-(3-Chloropropyl)piperidine 95%

Supplier: Apollo Scientific

OR950692-1G OR950692-250MG
APOSOR950692-1GEA 209 EUR
APOSOR950692-1G APOSOR950692-250MG
1-(3-Chloropropyl)piperidine 95%
1-(3-Chloropropyl)piperidine
Formula: C₈H₁₆ClN
MW: 161,672 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00044515
CAS Number: 1458-63-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR