Methyl 4-pyridylacetate hydrochloride 95%

Supplier: Apollo Scientific

OR951032-1G OR951032-25G OR951032-5G
APOSOR951032-1GEA 161 EUR
APOSOR951032-1G APOSOR951032-25G APOSOR951032-5G
Methyl 4-pyridylacetate hydrochloride 95%
Methyl 4-pyridylacetate hydrochloride
Formula: C₈H₉NO₂·ClH
Storage Temperature: Ambient
MDL Number: MFCD00236622
CAS Number: 70199-68-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR