1-(2-Methylphenyl)ethanamine 98%

Supplier: Apollo Scientific
Warning

OR951039-1G OR951039-25G OR951039-5G
APOSOR951039-1GEA 56.7 EUR
APOSOR951039-1G APOSOR951039-25G APOSOR951039-5G
1-(2-Methylphenyl)ethanamine 98%
1-(2-Methylphenyl)ethanamine
Formula: C₉H₁₃N
MW: 135,206 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05215232
CAS Number: 42142-17-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR