(1-Methyl-1H-benzoimidazol-2-yl)acetonitrile 95%

Supplier: Apollo Scientific

OR951177-1G OR951177-250MG
APOSOR951177-1GEA 669 EUR
APOSOR951177-1G APOSOR951177-250MG
(1-Methyl-1H-benzoimidazol-2-yl)acetonitrile 95%
(1-Methyl-1H-benzoimidazol-2-yl)acetonitrile
Formula: C₁₀H₉N₃
MW: 171,199 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00227239
CAS Number: 2735-62-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR