N-Ethyl-4-chlorobenzylamine 95%

Supplier: Apollo Scientific

OR951397-1G OR951397-5G
APOSOR951397-1GEA 125 EUR
APOSOR951397-1G APOSOR951397-5G
N-Ethyl-4-chlorobenzylamine 95%
N-Ethyl-4-chlorobenzylamine
Formula: C₉H₁₂ClN
MW: 169,651 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00045212
CAS Number: 69957-83-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR