2-Amino-6-chloroimidazo[1,2-b]pyridazine 95%
Supplier: Apollo Scientific
OR951417-1G
OR951417-250MG
APOSOR951417-1GEA
94.4
EUR
APOSOR951417-1G
APOSOR951417-250MG
2-Amino-6-chloroimidazo[1,2-b]pyridazine 95%
2-Amino-6-chloroimidazo[1,2-b]pyridazine
Formula:
C₆H₅ClN₄ Storage Temperature: Ambient |
MDL Number:
MFCD16495902 CAS Number: 887625-09-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...