3-(3-Thienyl)propanoic acid 95%

Supplier: Apollo Scientific

Synonyms: 3-(Thiophen-3-yl)propanoic acid

OR951642-1G OR951642-250MG
APOSOR951642-1GEA 102 EUR
APOSOR951642-1G APOSOR951642-250MG
3-(3-Thienyl)propanoic acid 95%
3-(3-Thienyl)propanoic acid
Formula: C₇H₈O₂S
MW: 156,202 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06655385
CAS Number: 16378-06-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR