(4-Butylphenyl)acetic acid 98%
Supplier: Apollo Scientific
OR951889-1G
OR951889-250MG
OR951889-5G
APOSOR951889-1GEA
214
EUR
APOSOR951889-1G
APOSOR951889-250MG
APOSOR951889-5G
(4-Butylphenyl)acetic acid 98%
(4-Butylphenyl)acetic acid
Formula:
C₁₂H₁₆O₂ MW: 192.254 g/mol |
MDL Number:
MFCD00982001 CAS Number: 14377-19-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...