2-Ethynyl-cyclopentanol 95%

Supplier: Apollo Scientific

OR952229-100MG OR952229-250MG
APOSOR952229-100MGEA 290 EUR
APOSOR952229-100MG APOSOR952229-250MG
2-Ethynyl-cyclopentanol 95%
2-Ethynyl-cyclopentanol
Formula: C₇H₁₀O
Storage Temperature: Ambient
MDL Number: MFCD18375237
CAS Number: 22022-30-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR