6-Cyclobutyl-3(2H)-pyridazinone 95%

Supplier: Apollo Scientific

OR952321-1G OR952321-250MG
APOSOR952321-1GEA 864 EUR
APOSOR952321-1G APOSOR952321-250MG
6-Cyclobutyl-3(2H)-pyridazinone 95%
6-Cyclobutyl-3(2H)-pyridazinone
Formula: C₈H₁₀N₂O
MW: 150,178 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11520596
CAS Number: 1161737-37-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR