N-Cyclopropyl-3-nitropyridin-4-amine 98%
Supplier: Apollo Scientific
OR952455-1G
OR952455-5G
APOSOR952455-1GEA
179
EUR
APOSOR952455-1G
APOSOR952455-5G
N-Cyclopropyl-3-nitropyridin-4-amine 98%
N-Cyclopropyl-3-nitropyridin-4-amine
Formula:
C₈H₉N₃O₂ MW: 179.176 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11184481 CAS Number: 380605-28-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...