6-Pyridin-3-yl-2H-pyridazin-3-one 97%
Supplier: Apollo Scientific
OR952501-1G
OR952501-250MG
APOSOR952501-1GEA
633
EUR
APOSOR952501-1G
APOSOR952501-250MG
6-Pyridin-3-yl-2H-pyridazin-3-one 97%
6-Pyridin-3-yl-2H-pyridazin-3-one
Formula:
C₉H₇N₃O MW: 173,171 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09033893 CAS Number: 78784-65-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...