7-Amino-3,4-dihydroquinoxalin-2(1H)-one dihydrochloride 97%

Supplier: Apollo Scientific

OR952537-1G OR952537-5G
APOSOR952537-1GEA 139 EUR
APOSOR952537-1G APOSOR952537-5G
7-Amino-3,4-dihydroquinoxalin-2(1H)-one dihydrochloride 97%
7-Amino-3,4-dihydroquinoxalin-2(1H)-one dihydrochloride
Formula: C₈H₉N₃O·2ClH
MW: 236,098 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09878748
CAS Number: 860503-30-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR