7-Chloro-2,3,4,5-tetrahydro-1H-benzo[e][1,4]diazepine 97%
Supplier: Apollo Scientific
OR952589-1G
APOSOR952589-1GEA
121
EUR
APOSOR952589-1G
7-Chloro-2,3,4,5-tetrahydro-1H-benzo[e][1,4]diazepine 97%
7-Chloro-2,3,4,5-tetrahydro-1H-benzo[e][1,4]diazepine
Formula:
C₉H₁₁ClN₂ MW: 182,65 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07369930 CAS Number: 57756-37-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...