4-Methyl-2-(methylthio)-5-pyrimidinecarbonitrile 95%
Supplier: Apollo Scientific
OR952620-1G
OR952620-250MG
APOSOR952620-1GEA
719
EUR
APOSOR952620-1G
APOSOR952620-250MG
4-Methyl-2-(methylthio)-5-pyrimidinecarbonitrile 95%
4-Methyl-2-(methylthio)-5-pyrimidinecarbonitrile
Formula:
C₇H₇N₃S Storage Temperature: Ambient |
MDL Number:
MFCD14585003 CAS Number: 89079-62-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...