4-(Methylsulphonamido)benzaldehyde 95%

Supplier: Apollo Scientific

OR952701-1G OR952701-5G
APOSOR952701-1GEA 179 EUR
APOSOR952701-1G APOSOR952701-5G
4-(Methylsulphonamido)benzaldehyde 95%
4-(Methylsulphonamido)benzaldehyde
Formula: C₈H₉NO₃S
Storage Temperature: Ambient
MDL Number: MFCD11052340
CAS Number: 83922-54-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR