8-Methoxy-2,3,4,5-tetrahydro-benzo[c]azepin-1-one 95%
Supplier: Apollo Scientific
OR952737-1G
OR952737-250MG
APOSOR952737-1GEA
720
EUR
APOSOR952737-1G
APOSOR952737-250MG
8-Methoxy-2,3,4,5-tetrahydro-benzo[c]azepin-1-one 95%
8-Methoxy-2,3,4,5-tetrahydro-benzo[c]azepin-1-one
Formula:
C₁₁H₁₃NO₂ MW: 191,226 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD02063284 CAS Number: 22246-71-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...