3-(Chloromethyl)pyrazole hydrochloride 96%
Supplier: Apollo Scientific
OR952758-1G
OR952758-5G
APOSOR952758-1GEA
112
EUR
APOSOR952758-1G
APOSOR952758-5G
3-(Chloromethyl)pyrazole hydrochloride 96%
3-(Chloromethyl)pyrazole hydrochloride
Formula:
C₄H₅ClN₂·ClH Storage Temperature: Ambient |
MDL Number:
MFCD08234808 CAS Number: 69658-97-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...