7-Methyl-1H-indazol-5-ol 95%
Supplier: Apollo Scientific
Synonyms:
5-Hydroxy-7-methyl-1H-indazole
OR952761-1G
OR952761-250MG
APOSOR952761-1GEA
193
EUR
APOSOR952761-1G
APOSOR952761-250MG
7-Methyl-1H-indazol-5-ol 95%
7-Methyl-1H-indazol-5-ol
Formula:
C₈H₈N₂O MW: 148,162 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD12198892 CAS Number: 478841-61-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...