9-Methyl-2,3,4,5-tetrahydro-1H-benzo[e][1,4]diazepine 95%
Supplier: Apollo Scientific
OR952781-1G
OR952781-250MG
APOSOR952781-1GEA
574
EUR
APOSOR952781-1G
APOSOR952781-250MG
9-Methyl-2,3,4,5-tetrahydro-1H-benzo[e][1,4]diazepine 95%
9-Methyl-2,3,4,5-tetrahydro-1H-benzo[e][1,4]diazepine
Formula:
C₁₀H₁₄N₂ MW: 162,232 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07369976 CAS Number: 195986-82-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...