2-(O-Tolylamino)ethanol 95%
Supplier: Apollo Scientific
OR952803-1G
OR952803-250MG
APOSOR952803-1GEA
109
EUR
APOSOR952803-1G
APOSOR952803-250MG
2-(O-Tolylamino)ethanol 95%
2-(O-Tolylamino)ethanol
Formula:
C₉H₁₃NO Storage Temperature: Ambient |
MDL Number:
MFCD00083105 CAS Number: 136-80-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...