1-(4-Aminophenyl)cyclopentanecarbonitrile 95%

Supplier: Apollo Scientific

OR952958-1G OR952958-250MG
APOSOR952958-1GEA 0 EUR
APOSOR952958-1G APOSOR952958-250MG
1-(4-Aminophenyl)cyclopentanecarbonitrile 95%
1-(4-Aminophenyl)cyclopentanecarbonitrile
Formula: C₁₂H₁₄N₂
Storage Temperature: Ambient
MDL Number: MFCD00807534
CAS Number: 115279-73-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR