Ethyl 2-hydroxy-4-methyl-5-pyrimidinecarboxylate 95%
Supplier: Apollo Scientific
Synonyms:
Ethyl 2-hydroxy-4-methylpyrimidine-5-carboxylate
OR953278-1G
OR953278-5G
APOSOR953278-1GEA
125
EUR
APOSOR953278-1G
APOSOR953278-5G
Ethyl 2-hydroxy-4-methyl-5-pyrimidinecarboxylate 95%
Ethyl 2-hydroxy-4-methyl-5-pyrimidinecarboxylate
Formula:
C₈H₁₀N₂O₃ MW: 182,177 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01875049 CAS Number: 6214-64-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...