Methyl acetimidate hydrochloride 90%

Supplier: Apollo Scientific

OR953398-100G OR953398-25G OR953398-5G OR953398-1G
APOSOR953398-100GEA 945 EUR
APOSOR953398-100G APOSOR953398-25G APOSOR953398-5G APOSOR953398-1G
Methyl acetimidate hydrochloride 90%
Methyl acetimidate hydrochloride
Formula: C₃H₇NO·ClH
MW: 109,555 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00012571
CAS Number: 14777-27-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR