Methyl 3-(chlorosulphonyl)propanoate 97%
|
Danger
|
OR953596-10G
OR953596-1G
OR953596-5G
APOSOR953596-10GEA
173
EUR
APOSOR953596-10G
APOSOR953596-1G
APOSOR953596-5G
Methyl 3-(chlorosulphonyl)propanoate 97%
Methyl 3-(chlorosulphonyl)propanoate
Formula:
C₄H₇ClO₄S MW: 186,614 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07339056 CAS Number: 15441-07-3 UN: 3265 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...