2,5-Dimethyl-1H-pyrrole-3-carbaldehyde 95%

Supplier: Apollo Scientific

OR953835-1G OR953835-250MG OR953835-5G
APOSOR953835-1GEA 248 EUR
APOSOR953835-1G APOSOR953835-250MG APOSOR953835-5G
2,5-Dimethyl-1H-pyrrole-3-carbaldehyde 95%
2,5-Dimethyl-1H-pyrrole-3-carbaldehyde
Formula: C₇H₉NO
MW: 123,152 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01830401
CAS Number: 2199-63-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR