4H-3,1-Benzothiazine-2-thiol 98%

Supplier: Apollo Scientific

OR953867-1G OR953867-250MG
APOSOR953867-1GEA 435 EUR
APOSOR953867-1G APOSOR953867-250MG
4H-3,1-Benzothiazine-2-thiol 98%
4H-3,1-Benzothiazine-2-thiol
Formula: C₈H₇NS₂
Storage Temperature: Ambient
MDL Number: MFCD07339560
CAS Number: 493-64-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR