3-Isobutoxy propylamine 98%

Supplier: Apollo Scientific

OR953927-1G OR953927-250MG OR953927-5G
APOSOR953927-1GEA 292 EUR
APOSOR953927-1G APOSOR953927-250MG APOSOR953927-5G
3-Isobutoxy propylamine 98%
3-Isobutoxy propylamine
Formula: C₇H₁₇NO
MW: 131,216 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00025618
CAS Number: 32923-88-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR