1-Ethyl-1,3-benzodiazole-5-carbonitrile 96%
Supplier: Apollo Scientific
OR954285-1G
OR954285-5G
APOSOR954285-1GEA
183
EUR
APOSOR954285-1G
APOSOR954285-5G
1-Ethyl-1,3-benzodiazole-5-carbonitrile 96%
1-Ethyl-1,3-benzodiazole-5-carbonitrile
Formula:
C₁₀H₉N₃ MW: 171,199 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD12827521 CAS Number: 1120244-47-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...