4-(3-Hydroxyphenyl)phenol 98%
Supplier: Apollo Scientific
OR954776-1G
OR954776-5G
APOSOR954776-1GEA
219
EUR
APOSOR954776-1G
APOSOR954776-5G
4-(3-Hydroxyphenyl)phenol 98%
4-(3-Hydroxyphenyl)phenol
Formula:
C₁₂H₁₀O₂ MW: 186.207 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11895234 CAS Number: 18855-13-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...