(3,5-Dimethylpyridin-2-yl)methanol 95%

Supplier: Apollo Scientific

OR955234-1G OR955234-250MG
APOSOR955234-1GEA 379 EUR
APOSOR955234-1G APOSOR955234-250MG
(3,5-Dimethylpyridin-2-yl)methanol 95%
(3,5-Dimethylpyridin-2-yl)methanol
Formula: C₈H₁₁NO
MW: 137,179 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09839185
CAS Number: 202932-05-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR