3-(4-Hydroxyphenyl)pyridine-2-carbonitrile 96%

Supplier: Apollo Scientific

OR955386-1G OR955386-250MG
APOSOR955386-1GEA 754 EUR
APOSOR955386-1G APOSOR955386-250MG
3-(4-Hydroxyphenyl)pyridine-2-carbonitrile 96%
3-(4-Hydroxyphenyl)pyridine-2-carbonitrile
Formula: C₁₂H₈N₂O
Storage Temperature: Ambient
MDL Number: MFCD22689141
CAS Number: 1235035-68-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR