Methyl 3-(2-chlorophenyl)propanoate 95%
Supplier: Apollo Scientific
OR955438-1G
OR955438-100MG
OR955438-250MG
OR955438-500MG
APOSOR955438-1GEA
633
EUR
APOSOR955438-1G
APOSOR955438-100MG
APOSOR955438-250MG
APOSOR955438-500MG
Methyl 3-(2-chlorophenyl)propanoate 95%
Methyl 3-(2-chlorophenyl)propanoate
Formula:
C₁₀H₁₁ClO₂ MW: 198,646 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD16620959 CAS Number: 74124-86-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...