Methyl 3,5-dimethylisoxazole-4-carboxylate 95%

Supplier: Apollo Scientific

OR955579-1G OR955579-5G
APOSOR955579-1GEA 212 EUR
APOSOR955579-1G APOSOR955579-5G
Methyl 3,5-dimethylisoxazole-4-carboxylate 95%
Methyl 3,5-dimethylisoxazole-4-carboxylate
Formula: C₇H₉NO₃
MW: 155,151 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06203795
CAS Number: 56328-87-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR