Methyl 5,6,7,8-tetrahydronaphthalene-1-carboxylate 95%
Supplier: Apollo Scientific
OR955660-1G
APOSOR955660-1GEA
223
EUR
APOSOR955660-1G
Methyl 5,6,7,8-tetrahydronaphthalene-1-carboxylate 95%
Methyl 5,6,7,8-tetrahydronaphthalene-1-carboxylate
Formula:
C₁₂H₁₄O₂ MW: 190,238 g/mol |
MDL Number:
MFCD15523539 CAS Number: 66193-59-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...